Order Entry
Austria
ContactUsLinkComponent
Naphthol AS-BI phosphate solution 12.5 mg/mL, Sigma-Aldrich®
Naphthol AS-BI phosphate solution 12.5 mg/mL, Sigma-Aldrich®
Catalog # 3871-10ML
CAS Number:  
Naphthol AS-BI phosphate solution 12.5 mg/mL, Sigma-Aldrich®
Catalog # 3871-10ML
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :452.1926 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
Storage Temperature:Refrigerator
 :1
CAS Number:1919-91-1
Formula:C₁₈H₁₅BrNO₆P
MDL Number:MFCD00004058