Order Entry
Austria
ContactUsLinkComponent
Irritant
Health
Fetal Hemoglobin Kit, Sigma-Aldrich®
Catalog # 285C-1KT
CAS Number:  
Irritant
Health
Fetal Hemoglobin Kit, Sigma-Aldrich®
Catalog # 285C-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ600 (more info.)
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
 :1