




Sudan Black B Staining System, Sigma-Aldrich®
Catalog # 380B-1KT
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
About this item
:
:M-Clarity™ quality level = MQ500 (more info.)
UN:3316
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
:1