ContactUsLinkComponent
Molecular detection assay, Salmonella
Molecular detection assay, Salmonella
Catalog # MMMPMDA2SAL96
Supplier:  3M Food Safety
undefined
Molecular detection assay, Salmonella
Catalog # MMMPMDA2SAL96
Supplier:  3M Food Safety
Specifications
  • Description:
    Salmonella 96 tests/kit
  • Pk:
    96 Tests
Specifications
About this item
3M™ Molecular Detection Assay 2 - Salmonella is used with the 3M™ Molecular Detection System for the rapid and specific detection of Salmonella species in enriched food and environmental samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster
As with all test methods, the source, formulation and quality of enrichment medium can influence the results. 3M has evaluated the 3M Molecular Detection Assay 2 Salmonella with Buffered Peptone Water ISO.