ContactUsLinkComponent
Water positive control (30 ng), HWATP10, Clean-Trace™
Water positive control (30 ng), HWATP10, Clean-Trace™
Catalog # MMMPHWATP10
Supplier:  3M Food Safety
undefined
Water positive control (30 ng), HWATP10, Clean-Trace™
Catalog # MMMPHWATP10
Supplier:  3M Food Safety
Specifications
  • Description:
    Clean-Trace™ water positive control (30 ng)
  • Seq:
    18
  • Pk:
    10
Specifications
About this item
Designed to provide assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly.
  • Verifies the performance of reagents and instruments
  • Can be used as a quench test with 3M™ Clean-Trace™ Water ATP tests
Designed to enhance your hygiene monitoring procedures by providing assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly to ensure reliable results.