Order Entry
Austria
ContactUsLinkComponent
Neutral mounting medium, DPX, Sigma-Aldrich®
Neutral mounting medium, DPX, Sigma-Aldrich®
Catalog # 317616-100ML
CAS Number:  
Neutral mounting medium, DPX, Sigma-Aldrich®
Catalog # 317616-100ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Neutral mounting medium, DPX
  • MDL No.:
    MFCD00135554

Specifications

About this item

D.P.X mounting medium is a mixture of distyrene (polystyrene), a plasticizer (tricresyl phosphate), and xylene. It facilitates the protection of stains and can dry quickly.

Caution: Storage temperature: Room temperature