Order Entry
Austria
ContactUsLinkComponent
Reticulum Stain Kit acc. to Gordon & Sweets, Sigma-Aldrich®
Reticulum Stain Kit acc. to Gordon & Sweets, Sigma-Aldrich®
Catalog # HT102A-1KT
CAS Number:  
undefined
Reticulum Stain Kit acc. to Gordon & Sweets, Sigma-Aldrich®
Catalog # HT102A-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
UN:3316
 :GHS05
 :GHS09
Storage Temperature:Refrigerator
 :1