Order Entry
Austria
ContactUsLinkComponent
Immersion oil viscosity 150 cSt (lit.), Sigma-Aldrich®
Immersion oil viscosity 150 cSt (lit.), Sigma-Aldrich®
Certificates
Immersion oil viscosity 150 cSt (lit.), Sigma-Aldrich®

Specifications

  • Form:
    Liquid
  • M-Clarity quality level:
    MQ200
  • Viscosity:
    150 cSt(lit.)
  • Storage temperature:
    RT
  • Density:
    1.080 g/cm³

Specifications

Frequently Bought Together