Order Entry
Austria
ContactUsLinkComponent
α-Naphthyl Butyrate Esterase, Lymphocyte kit, Sigma-Aldrich®
α-Naphthyl Butyrate Esterase, Lymphocyte kit, Sigma-Aldrich®
Catalog # 181B-1KT
CAS Number:  
α-Naphthyl Butyrate Esterase, Lymphocyte kit, Sigma-Aldrich®
Catalog # 181B-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ500 (more info.)
UN:3316
 :GHS02
 :GHS03
 :GHS05
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Refrigerator
 :1