Immersion oil for microscopy, Sigma-Aldrich®
Immersion oil is used for high resolution (1000X) light microscopy work under oil immersion objective lens.
- Suitable for Hematology
- Suitable for Histology
- MDL No.:MFCD00131369
- M-Clarity quality level:MQ100
- Viscosity:100-120 mPa.s (20 °C)
- Refractive index:n20/D 1.516
- Density:1.025 g/mL (20 °C)
Frequently Bought Together