Order Entry
Austria
ContactUsLinkComponent
 
VWR®, Fast HiFi DNA Polymerase, 2 U/µl
 
undefined
VWR®, Fast HiFi DNA Polymerase, 2 U/µl

 

  • Application:
    Cloning (high fidelity)
    Standard amplification
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    Yes
  • Hot start:
    No
  • Storage conditions:
    –20 °C (long-term), +4 °C (6 months)
  • Amplicon size:
    18 kb for gDNA
  • Processivity:
    Fast
  • Exonuclease activity:
    5’→3’ 3’→5’
  • Product overhang:
    No
  • Features:
    feature-ls-33964141|feature-ls-33964143
  • Buffer composition:
    5X buffer with 7,5 mM MgCl₂
  • Concentration:
    2 U/µl
  • Description:
    Fast HiFi DNA Polymerase 2 U/µl 2500 units
  • Size:
    5×0,25 ml + 14×1,5 ml
  • Units:
    2500

 

Frequently Bought Together