ContactUsLinkComponent
Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Artikelnr. ADLM6031535
undefined
Spare LED tubes for Apollo II and Apollo II high intensity liquid viewers
Artikelnr. ADLM6031535
Specifications
  • Description:
    Apollo II LED tubes - T5
  • Pk:
    2
Specifications
About this item
Pack of two 10 Watt LED bulbs (550 mm long) with a diffuser, which give a light level of between 2000 and 3750 Lux. The benefit of using LED tubes is more consistent light over the years compared with fluorescent tubes. These LED bulbs have a service life of 50,000 hours.
  • Colour temperature: 4000 K
  • Colour Rendering Index (CRI) Ra: greater than 80
  • Luminous flux at 25 °C: 950 - 1150 Im
The same bulbs to be used for Apollo II (for transparent glass) as well as Apollo II High Intensity (for colour glass).