ContactUsLinkComponent
Hand towel, interfolded, SCOTT® ESSENTIAL™
Hand towel, interfolded, SCOTT® ESSENTIAL™
Catalog # KIMB6617
Supplier:  KIMBERLY CLARK
undefined
Hand towel, interfolded, SCOTT® ESSENTIAL™
Catalog # KIMB6617
Supplier:  KIMBERLY CLARK
Specifications
  • Colour:
    White
  • Description:
    Hand towel small
  • L×W:
    210×200 mm
  • Pk:
    5.100
Specifications
About this item
Designed to help you support washroom hygiene standards in high frequency environments without compromising on quality and value.
  • Touch free dispensers reduce the risk of cross contamination
  • With 340 hand towels per clip, maintenance time is reduced
Interfold paper towels, or V fold paper towels are designed to dispense one paper hand towel at a time. This single sheet dispensing method is designed to improve hand hygiene by minimising the risk of contamination when hand drying as the user only touches the sheet they're using.
These folded paper hand towels also feature our unique Airflex™ Technology resulting in faster absorbency for more hygienic hand drying, whilst also using less and creating less waste, helping you get more for your spend whilst improving workplace hygiene.
Certifications: FSC and European Ecolabel certified for environmental assurance. Food contact certified.
Packaging: 340 towels per clip, 15 clips per case.
Dispenser
Product Information