Orders LinkContactUsLinkComponent
Neutralisation unit, SIMVA, behrotest®
Neutralisation unit, SIMVA, behrotest®
SIMVAC Neutralisation unit
Artikelnr. BEHRB00217922
Anbieter:   BEHR
Neutralisation unit, SIMVA, behrotest®
Artikelnr. BEHRB00217922
Anbieter:  BEHR
Neutralisation unit, SIMVA, behrotest®
Neutralisation unit, SIMVA, behrotest®
SIMVAC Neutralisation unit
Preis
494,98 €
Log in for availability
1 * 1 ST
Product Details & Documents
For behr digestion units (K-block & Inkjel devices), extraction systems for acid vapours are a necessity. We differentiate between a simple water jet pump / SIMVAC and the behrosog3 extraction system and neutraliser.
  • SIMVAC can be used for digestion blocks with up to 8 sample positions
  • Only requires a water connection to generate the required extraction performance – nothing else
  • Easy, robust and price effective
Specifications
  • Description:
    SIMVAC Neutralisation unit