Cleanroom face masks, Kimtech™ M5
Catalog # 111-0383
Supplier: KIMBERLY CLARK
Specifications
- Type:With earloops
- Material:Polyester/cellulose
- Colour:Blue
- Pk:500
Specifications
About this item
Kimtech™ Pure M5 Pleat-style face masks with ear loops are a smart piece of personal protective equipment (PPE).
- Three-layer design of polyester and cellulose, with a soft inner layer
- The pleat-style masks have ear loops and a fully enclosed bendable nosepiece
- Ultra sonically bonded ear loops are durable
They provide good breathability for comfort. These double bagged face masks are popular for a variety of environments, including those for biotech, medical devices, electronics and telecommunications.