Gloves, Latex, Kimberly-Clark™ Professional Kimtech Pure™
Supplier: KIMBERLY CLARK
Certificates
About this item
Natural rubber latex for tactile sensitivity and comfort.
- Textured palm and fingertips
- Beaded cuff
- Hand-specific
- Low residual protein content
- Double-bagged, multi-pack
Recommended for ISO Class 3 or higher cleanroom environments.
Frequently Bought Together