Sampling scoop, long handle, disposable Bio
Supplier: BURKLE
Certificates
About this item
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings. With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
- Product made from renewable raw materials
- Bio-PE, white
- Individually packaged and optionally sterilised
- Handle length 300 mm
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
The disposable scoop is suitable for sensitive areas of pharmaceuticals and chemistry as well as the food and cosmetics industries.
Specifications
- Catalog No:
- 331-0354
- 331-0355
- 331-0356
- 331-0357
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact, Sustainable Materials
- Environmental certification:
- ACT-Label
- ACT-Label
- ACT-Label
- ACT-Label