Sampling scoops, long handle, disposable, LaboPlast®/SteriPlast®
Supplier: BURKLE
Certificates
About this item
This sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content, or to reach through narrow openings.
- PS, white
- Individually packed and optionally sterilised
- Handle length: 300 mm
- For single use
- Cleanroom manufactured (clean room Class 7)
With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container. The disposable scoop is suitable for sensitive areas of pharmaceuticals and chemistry as well as the food and cosmetics industries.
Certifications: Complies with EU food requirements and FDA regulations.
Frequently Bought Together