Orders LinkContactUsLinkComponent
RackScan
RackScan
Supplier:   EPPENDORF
RackScan
Supplier:  EPPENDORF
Certificates
RackScan
2 Options Available Below
Product Details & Documents
The RackScan instrument can read up to 96 barcoded vials in parallel.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.
Product Family Options
Product Information
  • Availability
    Price
  • EPPE6002000.014
    RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
    RackScan s, code scanner for side reading of SBS racks and cryostorage boxes, accessory for the RackScan product family
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 570,00 €
    1 570,00 €
    1 * 1 ST
       1 570,00 €
  • EPPE6002000.006
    RackScan b, scanner (bottom reading) for 2D-coded tubes in SBS racks and cryostorage boxes, includes software RackScan analyser
    RackScan b, scanner (bottom reading) for 2D-coded tubes in SBS racks and cryostorage boxes, includes software RackScan analyser
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 720,00 €
    10 720,00 €
    1 * 1 ST
       10 720,00 €