Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Quick TB Kit
Quick TB Kit Cold Staining
Quick TB Kit Cold Staining
Quick TB Kit Cold Staining
Catalog # 720-1166
Supplier:
RAL
CAS Number:
Expand image
Quick TB Kit Cold Staining
Catalog # 720-1166
Supplier:
RAL
CAS Number:
Specifications
Pk:
1 KIT
Seq:
0075
Specifications
About this item
Product Details
Documentation
:
:
1
Product Details
Documentation