Order Entry
Austria
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99% (dried basis), HiPerSolv CHROMANORM® for ion pair chromatography
Sodium dodecyl sulphate (SDS) ≥99% (dried basis), HiPerSolv CHROMANORM® for ion pair chromatography
Catalog # 85798.130
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium dodecyl sulphate (SDS) ≥99% (dried basis), HiPerSolv CHROMANORM® for ion pair chromatography
Catalog # 85798.130
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    ≥ 99 %
  • Insoluble matter :
    Passes test
  • IR Spectrum:
    Passes test
  • pH (10 %) :
    6,0 - 7,5
  • Water :
    ≤ 2 %
  • Transmittance (200 nm) (0.005 mol/l) :
    ≥ 70 %
  • Transmittance (220 nm) (0.005 mol/l) :
    ≥ 90 %
  • Transmittance (250 nm) (0.005 mol/l) :
    ≥ 98 %

Specification Test Results

About this item

 :
 :288.38 g/mol
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175