Orders LinkContactUsLinkComponent
Core samplers
Core samplers
Supplier:   BURKLE
Core samplers
Supplier:  BURKLE
Certificates
Core samplers
3 Options Available Below
Product Details & Documents
These core samplers can be used to take samples of bulk materials, which can easily become stuck or settle during sampling due to their particle form.
  • Stainless steel AISI 316
  • For bulk materials that become stuck or settle easily
In comparison to standard all-layer samplers for bulk materials, the two-part core sampler has no gap dimensions in which the medium can become stuck while it is being removed. This makes it possible to take a proper sample of powders and granulates through multiple layers, such as glass balls or metal powders.
Depending on the size, the stainless steel sampler can be used for a wide range of different containers, for example big bags or small containers. The bulk material sampler takes a representative core sample and is ideal for use in various laboratory and industrial applications.
Delivery information: Delivery includes core sampler (consisting of boring rod and boring tube) and fixing clip.
Product Family Options
Product Information
  • Length
    Ø
    Volume
    Availability
    Price
  • 331-0374
    Core sampler
    Core sampler
    200 mm
    15 mm
    15 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 544,00 €
    544,00 €
    1 * 1 ST
       544,00 €
  • 331-0375
    Core sampler
    Core sampler
    400 mm
    25 mm
    110 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 592,00 €
    592,00 €
    1 * 1 ST
       592,00 €
  • 331-0376
    Core sampler
    Core sampler
    800 mm
    25 mm
    230 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 642,00 €
    642,00 €
    1 * 1 ST
       642,00 €