Plunger mat with adhesive backing, PIG®
Catalog # 115-2587
Supplier: NEW PIG
Specifications
- Type:Polypropylene
- Description:Plunger mats, 6 pads per box, 0,2 kg
- L×W:230×230 mm
- Pk:6
Specifications
About this item
Adhesive-backed mats keep the surfaces under plungers, brushes and soap dispensers neat and clean. Absorbent top soaks up water, urine and other liquids.
- Plunger mats control drips and eliminate puddles under toilet plungers and brushes, soap dispensers and other small-footprint areas in your restroom
- Permanently infused antimicrobial technology prevents the growth of odor-causing bacteria and mold on the mats to keep restrooms cleaner and fresher smelling
- Proprietary, full-coverage adhesive backing keeps mats in place underfoot with no shifting, curling or flipping up to stop slip and trip hazards; peels up easily
- Mats stick to your floor and won't budge while you clean them; simply mop them with the rest of the floor
- Durable, Hi-Trac™ surface stands up to daily cleaning and foot traffic
- Absorbs water and urine
EFM™ Absorbent technology captures water on contact, wicks it away and dries out quickly, Impermeable backing layers stop liquid pass-through to prevent stains and grout damage.
Release paper on the bottom of the mat should be removed prior to installation.
Certifications: Tested and certified by the National Floor Safety Institute (NFSI) as a high-traction surface which can reduce slip and fall claims by as much as 90% when used as part of a floor safety program.
Caution: Mats take on the properties of absorbed liquids - always take proper precautions when handling saturated absorbents.