Order Entry
Austria
ContactUsLinkComponent
Bradford's reagent for dye, ready-to-use
Corrosive
Irritant
Health
Bradford's reagent for dye, ready-to-use
Catalog # J61522.AP
CAS Number:  
Corrosive
Irritant
Health
Bradford's reagent for dye, ready-to-use
Catalog # J61522.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
UN:1805
ADR:8,III
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Ambient