Order Entry
Austria
Orders LinkContactUsLinkComponent
Ammonia solution standardised solution 0.1 N

Ammonia solution standardised solution 0.1 N

Glass bottle, 1 L
Catalog # 35576.K2
Ammonia solution standardised solution 0.1 N
Catalog # 35576.K2
CAS Number:  1336-21-6
Ammonia solution standardised solution 0.1 N
Ammonia solution standardised solution 0.1 N

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
66,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.