Disodium hydrogen phosphate, anhydrous 98.0-100.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
Artikelnr. CH05-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:PE pail, screw top
- Pk:12 kg
- Seq:4187
- Pk Info:1×12 kg
Specifications
Specification Test Results
- USP - Assay (Na₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Identification:Passes Test
- USP - Loss on Drying at 130°C:≤ 5.0 %
- USP - Insoluble Substances:≤ 0.4 %
- USP - Chloride (Cl):≤ 0.06 %
- USP - Sulfate (SO₄):≤ 0.2 %
- USP - Arsenic (As):≤ 16 ppm
- USP - Endotoxin Concentration (2.5 EU/g max.):Passes Test
- JPE - Assay (dried basis):≥ 99.0 %
- JPE - Arsenic (As):≤ 2 ppm
- JPE - Identification:Passes Test
- JPE - Carbonate:Passes Test
- JPE - Chloride (Cl):≤ 0.011 %
- JPE - Clarity and Color of Solution:Passes Test
- JPE - Heavy Metals:≤ 10 ppm
- JPE - pH (1-100):9.0 - 9.4
- JPE - Sulfate (SO₄):≤ 0.038 %
- JPE - Loss on Drying:≤ 2.0 %
- EP - Assay (Na₂HPO₄) (dried basis):98.0 - 101.0 %
- EP - Appearance of Solution:Passes Test
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP - Identification C:Passes Test
- EP - Identification D:Passes Test
- EP - Reducing Substances:Passes Test
- EP - Chloride (Cl):≤ 200 ppm
- EP - Iron (Fe):≤ 20 ppm
- EP - Loss on Drying:≤ 1.0 %
- EP - Monosodium Phosphate:≤ 2.5 %
- EP - Sulfate (SO₄):≤ 500 ppm
- ChP - Identification:Passes Test
- ChP - Alkalinity (pH):9.0 - 9.4
- ChP - Clarity and Color of Solution:Passes Test
- ChP - Chloride:≤ 0.001 %
- ChP - Sulfate:≤ 0.01 %
- ChP - Carbonate:Passes Test
- ChP - Water-insoluble substances:≤ 0.05 %
- ChP - Reducing substances:Passes Test
- ChP - Loss on drying @ 130 C:≤ 1.0 %
- ChP - Heavy metals:≤ 0.001 %
- ChP - Arsenic:≤ 0.0002 %
- ChP - Assay (dried basis):≥ 99.0 %
- ChP - Sodium dihydrogen phosphate:≤ 2.5 %
- ChP - Iron (Fe):≤ 0.002 %
Specification Test Results
About this item
:
:141.96 g/mol
:Management of Change (MOC) category = R
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496