Bottle, with GL 45 side neck, DURAN®
Catalog # SCOT219900701
Supplier: DWK Life Sciences
Specifications
- Cap Size:GL 45
- Ext.Ø:136 mm
- Capacity:2000 ml
- Height:260,6 mm
- Environmentally Preferable:
- Neck I-Ø:30 mm
- Pk:1
Specifications
About this item
HPLC 2000 ml reservoir bottle with two GL 45 inlets.
- Convenient – Side neck allow solvent filling without main delivery cap removal
- Safer – Capped bottles reduce the risk of volatile organic compounds (VOC) escape and spillage
- Compatible - GL 45 threads fit standard HPLC connection caps
- Manufactured from the material of choice for HPLC solvent bottles: borosilicate 3.3 glass offering a very low inorganic ion extractables profile
- Useful 2 L capacity
- Durable white enamel volume graduations (400 - 1800 ml, 200 ml intervals)
DURAN® bottles are ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents. Increasing the number of GL 45 inlets on the bottles adds convenience: one additional side neck allows additional solvent to be added without disturbing the main delivery cap.
Applications: mobile phase reservoirs for HPLC, reagent supply to liquid handling systems and analysers.
Certifications: Clear DURAN® is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a 'Type I, Class A low-expansion borosilicate glass' as defined by ASTM-E438-1992. Retrace Code, and certificate for manufacturing lot traceability.
Glass Type I according to the requirements of the current United States pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese pharmacopoeia JP <7.01> Test for Glass Containers for injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, the standard deviation of the batch and the date of issue.
Glass Type I according to the requirements of the current United States pharmacopoeia USP <660> Containers – Glass.
Glass Type I according to the requirements of the current European pharmacopoeia EP (3.2.1) Glass Containers for Pharmaceutical Use.
Glass Type I according to the requirements of the current Japanese pharmacopoeia JP <7.01> Test for Glass Containers for injections.
The calibration results of volumetric instruments with this certificate refer to one calibration batch.
This batch certificate confirms that ASTM Class A error limits required by USP are met. The certificate records the mean value, the standard deviation of the batch and the date of issue.