ContactUsLinkComponent
Reusable respirator filter retainer, Secure Click™
Reusable respirator filter retainer, Secure Click™
Catalog # 111-2566
Supplier:  3M
undefined
Reusable respirator filter retainer, Secure Click™
Catalog # 111-2566
Supplier:  3M
Specifications
  • Description:
    Secure Click™ reusable respirator filter retainer, D701
  • Seq:
    08
  • Pk:
    10
Specifications
About this item
Used to hold 3M Secure Click™ particulate filters D7N11 or D7P71 in place on Secure Click™ D8000 series cartridges.
  • Component works with Secure Click™ 800 series respirators and Secure Click™ D8000 series cartridges
  • Refer to the D7N11 or D7P71 filter 'User Instructions' for further information
The filter works with a 3M Secure Click™ 800 series respirators. Connection system works just like a seat belt. Simply align the connections and push until an audible click is heard. The filter is suitable for use in environments containing solid and liquid particles.