Order Entry
Austria
ContactUsLinkComponent
Triethylamine ≥99%, GPR RECTAPUR®
Triethylamine ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Triethylamine ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay :
    ≥ 99%
  • IR Spectrum:
    Passes test
  • Refractive index (20°C):
    1,399 - 1,401
  • Water:
    ≤ 0,2 %

Specification Test Results

Related Information

Frequently Bought Together