Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Potassium hydroxide 10% (w/v) in aqueous solution, StandARd®, Macron Fine Chemicals™
Catalog # MACRH320-01
CAS Number:  
Corrosive
Potassium hydroxide 10% (w/v) in aqueous solution, StandARd®, Macron Fine Chemicals™
Catalog # MACRH320-01
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay (KOH)(g/100 mL):
    9.5 - 10.5
  • Identification:
    Passes Test

Specification Test Results

About this item

 :
 :56.11 g/mol
ADR:8,II
 :Management of Change (MOC) category = N
EINECS:215-181-3
UN:1814
 :GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553