Orders LinkContactUsLinkComponent
Labels, polyester, Type B-494
Supplier:   Brady
Labels, polyester, Type B-494
Supplier:  Brady
Certificates
Labels, polyester, Type B-494
Labels, polyester, Type B-494
60 Options Available Below
Product Details & Documents
The B-494 polyester label is a reliable sample identification and traceability solution.
  • Use for cold temperature and cryogenic storage applications
  • Polyester material with acrylic adhesive and gloss finish
  • Maximum service temperature: 70 °C
  • Minimum service temperature: −196 °C
  • Label thickness is 0,0762 mm
  • Ribbon series R6400
The B-494 polyester label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen. Thanks to subsurface printing, the label's colour code will not fade, smear, or smudge, and will continue to offer fast sample recognition long after it was printed.
Die-cut labels are supplied on a roll, example 1×2". This node contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates. This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves.
Product Family Options
Product Information
  • Type
    Model
    Material
    Adhesive
    Colour
    Printing method
    Printer model
    Dimensions
    Availability
    Price
  • 116-0863
    Printer labels
    Printer labels
    B-494
    B33-250-494-BL
    Polyester
    Acrylic
    Blue/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0902
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0908
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0920
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0872
    Printer labels
    Printer labels
    B-494
    B33-310-494-BL
    Polyester
    Acrylic
    Blue/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0865
    Printer labels
    Printer labels
    B-494
    B33-250-494-GN
    Polyester
    Acrylic
    Green/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0901
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0907
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0919
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0871
    Printer labels
    Printer labels
    B-494
    B33-310-494-GN
    Polyester
    Acrylic
    Green/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0864
    Printer labels
    Printer labels
    B-494
    B33-250-494-OR
    Polyester
    Acrylic
    Orange/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0900
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0906
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0918
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0870
    Printer labels
    Printer labels
    B-494
    B33-310-494-OR
    Polyester
    Acrylic
    Orange/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0862
    Printer labels
    Printer labels
    B-494
    B33-250-494-RD
    Polyester
    Acrylic
    Red/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0898
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0904
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0916
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0868
    Printer labels
    Printer labels
    B-494
    B33-310-494-RD
    Polyester
    Acrylic
    Red/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0866
    Printer labels
    Printer labels
    B-494
    B33-250-494-PK
    Polyester
    Acrylic
    White/pink
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0899
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0905
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0917
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0869
    Printer labels
    Printer labels
    B-494
    B33-310-494-PK
    Polyester
    Acrylic
    White/pink
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0861
    Printer labels
    Printer labels
    B-494
    B33-250-494-YL
    Polyester
    Acrylic
    Yellow/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 152,00 €
    152,00 €
    1 * 1.500 ST
       152,00 €
  • 116-0897
    Printer labels
    Printer labels
    B-494
    THT-310-494-10-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 931,00 €
    931,00 €
    1 * 10.000 ST
       931,00 €
  • 116-0903
    Printer labels
    Printer labels
    B-494
    BPT-310-494-2.5-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 247,00 €
    247,00 €
    1 * 1 Rolle
       247,00 €
  • 116-0915
    Printer labels
    Printer labels
    B-494
    THT-306-494-3-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 330,00 €
    330,00 €
    1 * 1 Rolle
       330,00 €
  • 116-0867
    Printer labels
    Printer labels
    B-494
    B33-310-494-YL
    Polyester
    Acrylic
    Yellow/white
    BBP33
    12,7×25,4 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 252,00 €
    252,00 €
    1 * 2.500 ST
       252,00 €
  • 116-0884
    Printer labels
    Printer labels
    B-494
    B33-302-494-BL
    Polyester
    Acrylic
    Blue/white
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0883
    Printer labels
    Printer labels
    B-494
    B33-302-494-GN
    Polyester
    Acrylic
    Green/white
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0882
    Printer labels
    Printer labels
    B-494
    B33-302-494-OR
    Polyester
    Acrylic
    Orange/white
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0880
    Printer labels
    Printer labels
    B-494
    B33-302-494-RD
    Polyester
    Acrylic
    Red/white
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0881
    Printer labels
    Printer labels
    B-494
    B33-302-494-PK
    Polyester
    Acrylic
    White/pink
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0879
    Printer labels
    Printer labels
    B-494
    B33-302-494-YL
    Polyester
    Acrylic
    Yellow/white
    BBP33
    15,24×41,28 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 212,00 €
    212,00 €
    1 * 1.500 ST
       212,00 €
  • 116-0887
    Printer labels
    Printer labels
    B-494
    THT-179-494-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0889
    Printer labels
    Printer labels
    B-494
    THT-179-494-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0888
    Printer labels
    Printer labels
    B-494
    THT-179-494-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0886
    Printer labels
    Printer labels
    B-494
    THT-179-494-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0890
    Printer labels
    Printer labels
    B-494
    THT-179-494-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0896
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0895
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0894
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0892
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0893
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0891
    Printer labels
    Printer labels
    B-494
    THT-303-494-3-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    25,4×48,26 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 416,00 €
    416,00 €
    1 * 1 Rolle
       416,00 €
  • 116-0885
    Printer labels
    Printer labels
    B-494
    THT-179-494-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    25,4×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 415,00 €
    415,00 €
    1 * 1 Rolle
       415,00 €
  • 116-0914
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-BL
    Polyester
    Acrylic
    Blue/white
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0878
    Printer labels
    Printer labels
    B-494
    B33-304-494-BL
    Polyester
    Acrylic
    Blue/white
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €
  • 116-0913
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-GN
    Polyester
    Acrylic
    Green/white
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0877
    Printer labels
    Printer labels
    B-494
    B33-304-494-GN
    Polyester
    Acrylic
    Green/white
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €
  • 116-0912
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-OR
    Polyester
    Acrylic
    Orange/white
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0876
    Printer labels
    Printer labels
    B-494
    B33-304-494-OR
    Polyester
    Acrylic
    Orange/white
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €
  • 116-0910
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-RD
    Polyester
    Acrylic
    Red/white
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0874
    Printer labels
    Printer labels
    B-494
    B33-304-494-RD
    Polyester
    Acrylic
    Red/white
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €
  • 116-0911
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-PK
    Polyester
    Acrylic
    White/pink
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0875
    Printer labels
    Printer labels
    B-494
    B33-304-494-PK
    Polyester
    Acrylic
    White/pink
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €
  • 116-0909
    Printer labels
    Printer labels
    B-494
    THT-304-494-3-YL
    Polyester
    Acrylic
    Yellow/white
    Thermal transfer
    Thermal transfer
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Rolle
    1 * 1 Rolle - 201,00 €
    201,00 €
    1 * 1 Rolle
       201,00 €
  • 116-0873
    Printer labels
    Printer labels
    B-494
    B33-304-494-YL
    Polyester
    Acrylic
    Yellow/white
    BBP33
    9,53×25,4 mm
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 108,00 €
    108,00 €
    1 * 1.500 ST
       108,00 €