ContactUsLinkComponent
Acrylic Imhoff cone rack, Nalgene®
  532-0100
   Low Manufacturing Impact
undefined
Acrylic Imhoff cone rack, Nalgene®
  532-0100
   Low Manufacturing Impact
  • Description:
    Support for 3 Imhoff cones
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Securely hold three Imhoff cones with the durable and transparent Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack, featuring a wide base for stability on the lab bench.
  • Transparent; easily see level of contents
  • Can also be used to hold 500 and 1000 ml Nalgene separatory funnels