ContactUsLinkComponent
Cutting tool, WebSeal™, chromacol
Cutting tool, WebSeal™, chromacol
Catalog # 548-0233
Supplier:  CHROMACOL
Cutting tool, WebSeal™, chromacol
Catalog # 548-0233
Supplier:  CHROMACOL
Specifications
  • Description:
    WebSeal™ cutting tool
  • Seq:
    40
  • Pk:
    1
Specifications
About this item
This cutting tool allows removal of individual vials as a sealed unit, if required.