Order Entry
Austria
ContactUsLinkComponent
VWR® Face Masks, 2 ply, Non Woven PP
  113-0713
undefined
VWR® Face Masks, 2 ply, Non Woven PP
  113-0713

 

  • Type:
    With ear loops
  • Colour:
    White
  • Pk:
    50

 

 

This face mask, with nose bridge, is made from two ply, non woven PP.

  • Latex-free
  • Available with ear loops or ties
  • For industrial and cleanroom use only; do not use for protection against COVID-19
  • This product is not a medical device for medical or clinical use within the meaning of Regulation (EU) 2017/745 / Directive 93/42/EEC
  • This product is not a personal protective equipment (PPE) within the meaning of Regulation (EU) 2016/425

Density: 20 g/m² (front layer); 25 g/m² (back layer).