Safety boots, lace-up, Compositelite™ Traction, FD01
Supplier: Portwest
Certificates
About this item
Anti-static boots made from full-grain leather plus a dual-density PU/rubber sole, resistant to heat (300 °C).
- Composite toecap for added protection
- Pierce resistant composite midsole
- Energy absorbing seat region
- 100% waterproof membrane
- Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
- Fuel and oil resistant outsole
Combining comfort, ankle support, unrivalled anti slip properties and the finest materials and components, these boots are revelation in footwear construction. Lightweight construction with fastening zip, S3 water resistant leather and a waterproof and breathable membrane to keep feet dry in the harshest of conditions.
Certifications: EN ISO 20345:2011, S3, HRO, SRC; ASTM F2413-11; EN 61340-4-3 Class 3.
Frequently Bought Together