Flame retardant overalls, BIZ7, Bizflame™
: Portwest
The Hi vis anti-static Bizweld coverall offers all the benefits of flame resistant fabric with added hi vis and anti-static protection.
- Knee pad pockets
- Phone pocket
- Radio loops
- Flame resistant reflective tape
- Concealed stud front
Weight: 330 g/m²
: EN ISO 11612 (A1+A2, B1, C1, E3, F1), EN ISO 11611 Class 1 (A1+A2), EN 1149, EN ISO 20471 Class 3
- Catalog No:
- PRTWBIZ7YNR4XL
- PRTWBIZ7YNRXXXL
- PRTWBIZ7YNRL
- PRTWBIZ7YNRM
- PRTWBIZ7YNRXXL
- PRTWBIZ7YNRXL
- PRTWBIZ7YNRS
- Model:
- BIZ7
- BIZ7
- BIZ7
- BIZ7
- BIZ7
- BIZ7
- BIZ7
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Heat, Flame-retardant, Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- Bizflame™ fabric
- Bizflame™ fabric
- Bizflame™ fabric
- Bizflame™ fabric
- Bizflame™ fabric
- Bizflame™ fabric
- Bizflame™ fabric
- Weight:
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
- 330 g/m²
Frequently Bought Together