Order Entry
Austria
Orders LinkContactUsLinkComponent
ASPEC® 274 system

ASPEC® 274 system

GX-274 ASPEC, with verity 4260 duals, standard volume
Catalog # GILI2614010
Supplier:   GILSON LCMS
ASPEC® 274 system
Catalog # GILI2614010
Supplier:  GILSON LCMS
ASPEC® 274 system
ASPEC® 274 system

GX-274 ASPEC, with verity 4260 duals, standard volume

Price
32 404,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This ASPEC® system for positive pressure solid phase extraction (SPE) automation maximises throughput by processing up to four samples in parallel. With its small footprint and multiple probes, it is the ideal choice for reproducible and efficient sample cleanup for clinical, forensic, doping, food, and environmental testing labs.

  • Parallel processing for maximum efficiency
  • Positive pressure for reproducible extractions
  • Smart design for maximised performance
  • Intuitive error handling for trusted results
  • Powerful software control for optimised productivity
  • Versatile set-up for trace analysis

The ASPEC 274 system provides:
Four-probe Z-arm enables processing of four samples in parallel, increasing throughput and efficiency. Positive pressure extraction with unique sealing cap design increases reproducibility from sample to sample. The syringe pump delivers accurate volume dispensing and full control of sorbent drying. Large bed capacity for processing up to 144 samples unattended. The mobile racks accommodate 1, 3 and 6 ml SPE polypropylene cartridges, and enable multiple fraction elutions per sample. The syringe pump manages volumes from 1 to 10 ml with high accuracy and precision, allowing the transfer of eluted fractions in sealed injection vials or directly in the LC system with the optional GX direct injection module. The syringe pump pressure-sensing module provides error handling for high cartridge pressure or clogged liquid lines and monitors pressure on each fluid path individually. Optional jet wash rinse stations minimise carryover and cross contamination.

Specifications
  • Description:
    GX-274 ASPEC, with verity 4260 duals, standard volume
Complementary Products

Product Information

  • Catalog #
    Description
    Availability
    Price
  • GILI21063024
    TRILUTION® LH V4 software
    Log in for availability
    1 * 1 ST - 7 041,00 €

Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Rack, code 338, holds 64 12×32 mm vials (2 ml)
    Rack, code 338, holds 64 12×32 mm vials (2 ml)
    GILI260440106
    Log in for availability
    1 * 1 ST - 405,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Rack, code 343, holds 80 13×100 mm tubes
    Rack, code 343, holds 80 13×100 mm tubes
    GILI260440025
    Log in for availability
    1 * 1 ST - 426,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Rack, code 346, holds 44 16×100 mm tubes
    Rack, code 346, holds 44 16×100 mm tubes
    GILI260440049
    Log in for availability
    1 * 1 ST - 387,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Pressure regulator assay, 0 - 30 psi, ASPEC
    Pressure regulator assay, 0 - 30 psi, ASPEC
    GILI25051376
    Log in for availability
    1 * 1 ST - 556,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Syringe, 10 ml, 215/235
    Syringe, 10 ml, 215/235
    GILI25025345
    Log in for availability
    1 * 1 ST - 655,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Tubes glass, 15×85 mm, 10 ml, 250/pack
    Tubes glass, 15×85 mm, 10 ml, 250/pack
    GILI2954729
    Log in for availability
    1 * 250 Vial - 202,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Probe, 221×2,0×0,8 mm, septum piercing, beveled, grooved, 1,5 mm tip
    Probe, 221×2,0×0,8 mm, septum piercing, beveled, grooved, 1,5 mm tip
    GILI27067382
    Log in for availability
    1 * 1 ST - 258,67 €

Product Information

  • Catalog #
    Availability
    Price
  • Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)
    Rack, code 335, holds 48 15×45 mm wisp vials (4 ml)
    GILI260440083
    Log in for availability
    1 * 1 ST - 405,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Rack, code 341, holds 108 10×75 mm tubes
    Rack, code 341, holds 108 10×75 mm tubes
    GILI260440039
    Log in for availability
    1 * 1 ST - 381,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Tubes glass, 12×75 mm, 6 ml, 250/pack
    Tubes glass, 12×75 mm, 6 ml, 250/pack
    GILI2954726
    Log in for availability
    1 * 250 Vial - 102,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DEC accessory kit, ASPEC 1 ml GX27X
    DEC accessory kit, ASPEC 1 ml GX27X
    GILI2604701
    Log in for availability
    1 * 1 SET - 4 000,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DEC accessory kit, ASPEC 3 ml GX27X
    DEC accessory kit, ASPEC 3 ml GX27X
    GILI2604702
    Log in for availability
    1 * 1 SET - 4 000,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DEC accessory kit, ASPEC 6 ml GX27X
    DEC accessory kit, ASPEC 6 ml GX27X
    GILI2604704
    Log in for availability
    1 * 1 SET - 4 038,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Four probe rinse station or reservoir
    Four probe rinse station or reservoir
    GILI260440002
    Log in for availability
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.