ContactUsLinkComponent
Filter for half masks, P940
Filter for half masks, P940
Catalog # PRTWP940GRR
Supplier:  Portwest
Filter for half masks, P940
Catalog # PRTWP940GRR
Supplier:  Portwest
Specifications
  • Type:
    Particle filter, P940, grey
  • Pk:
    6
  • Class:
    P3
Specifications
About this item
P940 is a particle filter that protects against dusts. It is equipped with a special thread for pair use only and it can be used only on the P410 half mask.
  • Lightweight and comfortable
  • Screw-in connection
This filter is a P3 R type and protects against toxic substances such as particles and microorganisms.
Certifications: EN 143 Type P 3R