ContactUsLinkComponent
Fall protection harnesses, 2 point comfort
Fall protection harnesses, 2 point comfort
Catalog # PRTWFP14RER
Supplier:  Portwest
undefined
Fall protection harnesses, 2 point comfort
Catalog # PRTWFP14RER
Supplier:  Portwest
Specifications
  • Attachment Points:
    2-point
  • Description:
    Comfort harness
  • Pk:
    1
  • Weight:
    1,3 kg
Specifications
About this item
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Harness with a sliding dorsal D-Ring
  • Rear D-ring for keys or ID cards
  • Chest adjustment point
  • Padded belt
  • Made from polyester webbing, alloy steel, heat treated, polyester mesh
Features a breathable contoured backpad, two side D-Rings and flat bar buckles which are easy to use and fail safe. The waist, chest and leg adjusters offer excellent fit and user comfort.
Certifications: EN 361, EN 358