ContactUsLinkComponent
Ear plugs, PU foam, detectable, EP30
Catalog # PRTWEP30BLU
Supplier:  Portwest
undefined
Ear plugs, PU foam, detectable, EP30
Catalog # PRTWEP30BLU
Supplier:  Portwest
Specifications
  • Colour:
    Blue
  • Description:
    Ear plugs, corded
  • Environmentally Preferable:
  • Pk:
    200
Specifications
About this item
These detectable disposable ear plugs are comfortable to fit due to the soft PU foam material. Corded for increased safety and visibility in food industry environments. Each pair is inside a resealable hygienic polybag.
  • Can be used in food industries (metal detectable)
  • Retail box which aids presentation for retail sales
  • Individually packed for vending machines
These avoids the hearing problems occurring when working under hazardous conditions.
Certifications: EN 352-2 (SNR 37dB); ANSI S3.19 (32dB).
Packaging: 200/pack, 10/carton.