Cleanroom face masks, Auckland, P410
Lieferant: Portwest
Zertifikate
Über diesen Artikel
Economical half mask with low profile design providing a large field of vision.
- The soft adjustable strap ensures perfect fit to wide face contours
- Equipped with a dual (screw in) filter for enhanced breathing comfort
- Sweat port on the face piece for hot and humid environments
Manufactured from a durable yet soft non allergic TPE material which provides an excellent fit.
Certifications: EN 140
Lieferumfang: Filters sold separately.
Frequently Bought Together