Order Entry
Austria
ContactUsLinkComponent
Acetone ≥99.8% (by GC, corrected for water content), ChromAR® HPLC for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Irritant
Acetone ≥99.8% (by GC, corrected for water content), ChromAR® HPLC for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2435-25
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
Acetone ≥99.8% (by GC, corrected for water content), ChromAR® HPLC for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2435-25
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC, corrected for water):
    ≥ 99.8 %
  • Water (KF):
    ≤ 0.2 %
  • Titrable Acid (meq/g):
    ≤ 0.0005
  • Titrable Base (meq/g):
    ≤ 0.0006
  • Residue after Evaporation:
    ≤ 0.0005 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm:
    ≤ 0.8
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:
    ≤ 0.07
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤ 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 355 nm:
    ≤ 0.004
  • UV Cut-off:
    ≤ 327 nm

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765