Orders LinkContactUsLinkComponent
Darkfield optic ring for VisiLED slim ring light
Darkfield optic ring for VisiLED slim ring light
Darkfield optic ring for VisiLED slim ring light
Catalog # SCOC400810
Darkfield optic ring for VisiLED slim ring light
Catalog # SCOC400810
Darkfield optic ring for VisiLED slim ring light
Darkfield optic ring for VisiLED slim ring light
Darkfield optic ring for VisiLED slim ring light
Price
229,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Focus optic ring for darkfield illumination.
  • Focus optics ring for darkfield
  • Working distance: 5 to 25 mm
Designed for use only with VisiLED slim ring light (SCOC400150).
Ordering information: VisiLED slim ring light and controller MC 750, MC 1100, or MC 1500 required.
Documents
Specifications
  • Description:
    Darkfield optic ring for VisiLED slim ring light