Order Entry
Austria
ContactUsLinkComponent
Transmitted darkfield light stage, VisiLED, TLS-DF
Transmitted darkfield light stage, VisiLED, TLS-DF
Catalog # SCOC400350
undefined
Transmitted darkfield light stage, VisiLED, TLS-DF
Catalog # SCOC400350

Specifications

  • Description:
    VisiLED Transmitted light stage, int.Ø 50 mm (active), ext.Ø 140 mm
  • Pk:
    1

Specifications

About this item

Professional incident and darkfield illumination.

  • The VisiLED darkfield backlight TLS-DF offers the possibility of transmitted darkfield illumination
  • Lifetime is typically 50000 h by actively controlling the LED‘s temperature

Designed for use with controller MC 750, MC 1100, or MC 1500.

Ordering information: Required controllers must be purchased seperately.