Avantor® ACE® UltraCore BIO C4, Analytical HPLC/UHPLC Columns
: Avantor
Avantor® ACE® UltraCore BIO C4 are high performing solid-core (core-shell) particle columns, designed for a range of biomolecule applications. With both 300 Å and 500 Å pore sizes available, optimum pore size can be chosen. This phase is well suited to the analysis of hydrophobic proteins over 5 kDa molecular weight.
- Wide pore, solid-core combination for excellent peak shape & resolution of large molecules
- High purity, base deactivated silica with excellent reproducibility
- Butyl ligand well established in RP protein applications
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.
- Catalog No:
- BIO-351-0521
- BIO-251-1030
- BIO-251-0530
- BIO-351-546G
- BIO-251-1530
- BIO-251-521G
- BIO-351-0546
- BIO-351-1021
- BIO-251-0521
- BIO-251-546G
- BIO-351-1546
- BIO-351-1046
- BIO-351-1521
- BIO-251-1021
- BIO-351-0530
- BIO-351-530G
- BIO-251-0546
- BIO-351-1030
- BIO-251-1546
- BIO-251-1046
- BIO-251-1521
- BIO-251-530G
- BIO-351-1530
- BIO-351-521G
- Description:
- 300 Å
- 500 Å
- 500 Å
- For analytical columns with 4,6 mm Int.Ø
- 500 Å
- For analytical columns with 2,1 mm Int.Ø
- 300 Å
- 300 Å
- 500 Å
- For analytical columns with 4,6 mm Int.Ø
- 300 Å
- 300 Å
- 300 Å
- 500 Å
- 300 Å
- For analytical columns with 3,0 mm Int.Ø
- 500 Å
- 300 Å
- 500 Å
- 500 Å
- 500 Å
- For analytical columns with 3,0 mm Int.Ø
- 300 Å
- For analytical columns with 2,1 mm Int.Ø
- Carbon load:
- 0,4%
- 0,6%
- 0,6%
- 0,4%
- 0,6%
- 0,6%
- 0,4%
- 0,4%
- 0,6%
- 0,6%
- 0,4%
- 0,4%
- 0,4%
- 0,6%
- 0,4%
- 0,4%
- 0,6%
- 0,4%
- 0,6%
- 0,6%
- 0,6%
- 0,6%
- 0,4%
- 0,4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- 600 bar
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- 23 m²/g
- 16 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 23 m²/g
- 16 m²/g
- 16 m²/g
- Temperature limit:
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26