Thermoshaker with heating and cooling, Matrix Delta Plus
: IKA
The IKA Matrix Delta Plus has heating and cooling functionality and numerous user-friendly features.
- Large, clear display with a convenient menu structure allows for intuitive operation
- Programmable ramps allow for automatic execution of heating and heating tasks
- Secure, quick lock for fast and easy change of inserts and attachments as well as automatic attachment detection
- Aluminium die-cast housing ensures secure positioning, excellent stability and everyday robustness
- RS232 and USB interfaces
Matrix Delta Plus units heat and cool to ‒30 °C below ambient.
Whether it is blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays - samples with even the smallest volumes mix reliably and completely in all laboratory applications. No cross-contamination and an optimal mixing result.
: IP Protection Class according to DIN EN 60529: IP 21
: Supplied without accessories, which must be ordered separately. There is a wide range of accessories available for the IKA Matrix Delta Plus that allow the optimal operation for your specific application.
- Catalog No:
- 444-1323
- 444-1322
- Model:
- IKA Matrix Orbital Delta plus
- IKA Matrix Delta plus
- Incubating accuracy:
- ±1 at 20...45 °C
- ±1 at 20...45 °C
- Temperature control range:
- 15 below ambient…100 °C
- 30 below ambient…100 °C
- Heating speed:
- 18 °C/min
- 18 °C/min
- W×D×H:
- 216×147×351 mm
- 216×147×351 mm
- Weight:
- 9,2 kg
- 9,2 kg
- Orbit:
- 3 mm
- 3 mm
- Mixing speed range:
- 0 - 3000 min⁻¹
Frequently Bought Together