Order Entry
Austria
ContactUsLinkComponent
Disposable particle filtering respirators, FFP2, OP-Air Pro®
Disposable particle filtering respirators, FFP2, OP-Air Pro®
Catalog # HOPEM52210-WH50
Supplier:  Medicom
Disposable particle filtering respirators, FFP2, OP-Air Pro®
Catalog # HOPEM52210-WH50
Supplier:  Medicom

Specifications

  • Seq:
    04
  • Size:
    Small (225x90 mm)
  • Pk:
    50
  • With valve:
    -
  • Class:
    FFP2

Specifications

About this item

Designed to protect against high risk infectious diseases, these filtering half masks provide FFP2 NR D class protection against solid and liquid aerosols.

  • Category III and Medical Device Class I PPE
  • Bacterial Filtration Efficiency (BFE) is greater than 98%
  • Single-use, non sterile
  • Ultrasonic welding
  • Synthetic elastic headband
  • Latex-free

These folded masks feature a spunbond polypropylene inner/outer layer, with a melt-blown polypropylene filter. A polypropylene and iron nosepiece allows secure fitting. The masks provide 8 hours of effective protection against particulates and dust, and can also be used against non toxic solids and liquid aerosols.

Colour: White.

Certifications: EN 149:2001+A1:2009 FFP2 NR D. EN 14683:2014 Type IIR.