Analytical standard, Tetracyclin Hydrochlorid, reference material, Fluka™
Catalog # HONCR1773-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:64-75-5
- Description:Tetracyclin Hydrochlorid
- Formula:C₂₂H₂₄N₂O₈ · HCl
- Storage temperature:2…8 °C
- Description:Tetracyclin Hydrochlorid
- Form:Neat