Orders LinkContactUsLinkComponent
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Supplier:   Honeywell Chemicals
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
Certificates
Ionisation buffer solutions (CRMs), for AAS and ICP, Fluka™
3 Options Available Below
Product Details & Documents
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)
Delivery information: Supplied complete with a detailed Certificate of Analysis (CoA) in every delivery.
Product Family Options
Product Information
  • Pack type
    Pk
    Availability
    Price
  • HONCE4300-100ML
    2% Cesium (Cs) in 1% HNO₃
    2% Cesium (Cs) in 1% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 249,00 €
    249,00 €
    1 * 100 mL
       249,00 €
  • HONCE4301-100ML
    2% Lithium (Li) in 1% HNO₃
    2% Lithium (Li) in 1% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 249,00 €
    249,00 €
    1 * 100 mL
       249,00 €
  • HONCE4302-100ML
    2% Potassium (K) in 1% HCl 1%
    2% Potassium (K) in 1% HCl 1%
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 268,00 €
    268,00 €
    1 * 100 mL
       268,00 €