Order Entry
Austria
ContactUsLinkComponent
Tartrate Standard for Ion Chromatography, (CRM), Fluka™
Tartrate Standard for Ion Chromatography, (CRM), Fluka™
Catalog # HONCE3049-100ML
CAS Number:  
Tartrate Standard for Ion Chromatography, (CRM), Fluka™
Catalog # HONCE3049-100ML
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Tartrate standard for IC

Specifications

About this item

Certified Reference Material (CRM)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).